Skip to Content
Merck
All Photos(2)

Key Documents

227900

Sigma-Aldrich

Silver chlorate

99% trace metals basis

Synonym(s):

Argentous chlorate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$202.00

$202.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$202.00

About This Item

Linear Formula:
AgClO3
CAS Number:
Molecular Weight:
191.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$202.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99% trace metals basis

form

solid

impurities

≤1000.0 ppm Trace Metal Analysis

mp

230 °C (lit.)

density

4.43 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ag+].[O-]Cl(=O)=O

InChI

1S/Ag.ClHO3/c;2-1(3)4/h;(H,2,3,4)/q+1;/p-1

InChI key

SDLBJIZEEMKQKY-UHFFFAOYSA-M

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service