Skip to Content
Merck
All Photos(2)

Key Documents

227285

Sigma-Aldrich

1-Diazo-2-naphthol-4-sulfonic acid

powder

Synonym(s):

2,1-Naphthoquinone-1-diazide-4-sulfonic acid, 4-Diazo-3,4-dihydro-3-oxo-1-naphthalenesulfonic acid, 4-Sulfo-2,1-naphthoquinone diazide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$124.00

$124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$124.00

About This Item

Empirical Formula (Hill Notation):
C10H6N2O4S
CAS Number:
Molecular Weight:
250.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$124.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

contains

7% water

mp

160 °C (dec.) (lit.)

functional group

sulfonic acid

SMILES string

Oc1cc(c2ccccc2c1[N+]#N)S([O-])(=O)=O

InChI

1S/C10H6N2O4S/c11-12-10-7-4-2-1-3-6(7)9(5-8(10)13)17(14,15)16/h1-5H,(H-,13,14,15,16)

InChI key

QHIBNGIZPPHJAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The degradation and adsorption of 1-diazo-2-naphthol-4-sulfonic acid was studied using sewage sludge derived porous carbon[1].

Application

Involved in studies to analyze structure-activity relationships to predict biodegradability of N-heterocycles in wastewater and polluted soil

Reactant used as hydrazinonaphthalene and azonaphthalene thrombopoietin mimics acting as nonpeptidyl promoters of megakaryocytopoiesis

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lin Gu et al.
Journal of hazardous materials, 246-247, 145-153 (2013-01-10)
A sewage sludge derived porous carbon (SC), which was prepared by physicochemical activation and carbonization (600°C), was applied for the adsorption and degradation of 1-diazo-2-naphthol-4-sulfonic acid (1,2,4-Acid) in the presence of H(2)O(2) and the performance was compared to that of
Lin Gu et al.
Bioresource technology, 136, 719-724 (2013-04-10)
In this work, two types of sewage-sludges, aerobically digested and undigested sewage sludge, were used as precursors in the preparation of chars with magnetic property. The two solids were characterized to establish their textural and chemical properties. Due to different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service