Skip to Content
Merck
All Photos(1)

Key Documents

227153

Sigma-Aldrich

3-Chloro-2,4-pentanedione

97%

Synonym(s):

3-Chloroacetylacetone

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$180.00

$180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$180.00

About This Item

Linear Formula:
CH3COCHClCOCH3
CAS Number:
Molecular Weight:
134.56
Beilstein:
605870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$180.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.483 (lit.)

bp

49-52 °C/18 mmHg (lit.)

density

1.1921 g/mL at 20 °C (lit.)

functional group

chloro
ketone

storage temp.

2-8°C

SMILES string

CC(=O)C(Cl)C(C)=O

InChI

1S/C5H7ClO2/c1-3(7)5(6)4(2)8/h5H,1-2H3

InChI key

VLRGXXKFHVJQOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The tautomeric properties of 3-chloro-2,4-pentanedione were studied using gas electron diffraction (GED) and quantum chemical calculations[1].

Application

3-Chloro-2,4-pentanedione was used in the synthesis of tetrathiafulvenyl-acetylacetonate (TTFSacacH), precursor of novel redox-active ligands[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Massue et al.
Inorganic chemistry, 44(24), 8740-8748 (2005-11-22)
The reaction of tris(alkylthio)tetrathiafulvalene thiolates with 3-chloro-2,4-pentanedione affords tetrathiafulvalene (TTF) moieties substituted by the acetylacetone function (TTFSacacH), precursors of novel redox-active ligands: the acetylacetonate ions (TTFSacac). These TTFSacacHs have been characterized by X-ray diffraction analyses, and similar trends have been
Natalya V Belova et al.
The journal of physical chemistry. A, 112(14), 3209-3214 (2008-03-07)
The tautomeric properties of alpha-chlorinated acetylacetone, 3-chloro-2,4-pentanedione CH3C(O)-CHCl-C(O)CH3, have been investigated by gas electron diffraction (GED) and quantum chemical calculations (B3LYP and MP2 approximations with different basis sets up to cc-pVTZ). Analysis of the GED intensities resulted in the presence
Almeqdad Y Habashneh et al.
Archiv der Pharmazie, 347(6), 415-422 (2014-03-13)
A new series of N1-(flavon-6-yl)amidrazones were synthesized by reacting the hydrazonoyl chloride derived from 6-aminoflavone with the appropriate sec-cyclic amines. The antitumor activities of these compounds were evaluated on breast cancer (MCF-7) and leukemic (K562) cell lines. Among the compounds
Jennifer A Jacobsen et al.
Journal of medicinal chemistry, 54(2), 591-602 (2010-12-30)
Fragment-based lead design (FBLD) has been used to identify new metal-binding groups for metalloenzyme inhibitors. When screened at 1 mM, a chelator fragment library (CFL-1.1) of 96 compounds produced hit rates ranging from 29% to 43% for five matrix metalloproteases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service