Skip to Content
Merck
All Photos(1)

Key Documents

226254

Sigma-Aldrich

N,N-Diisopropylformamide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$82.20

$82.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$82.20

About This Item

Linear Formula:
HCON[CH(CH3)2]2
CAS Number:
Molecular Weight:
129.20
Beilstein:
1745406
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$82.20


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

196 °C (lit.)

mp

10-11 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

[H]C(=O)N(C(C)C)C(C)C

InChI

1S/C7H15NO/c1-6(2)8(5-9)7(3)4/h5-7H,1-4H3

InChI key

UNBDDZDKBWPHAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The chemical ionisation and electron ionisation mass spectroscopic data, retention indices and NMR spectra of N,N-diisopropylformamide was studied[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F B Hopkins et al.
Analytical and bioanalytical chemistry, 406(21), 5111-5119 (2014-03-19)
Detailed chemical analysis of solutions used to decontaminate chemical warfare agents can be used to support verification and forensic attribution. Decontamination solutions are amongst the most difficult matrices for chemical analysis because of their corrosive and potentially emulsion-based nature. Consequently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service