Skip to Content
Merck
All Photos(1)

Documents

226033

Sigma-Aldrich

1-Cyanovinyl acetate

98%

Synonym(s):

α-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, 2-Hydroxyacrylonitrile acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(OCOCH3)CN
CAS Number:
Molecular Weight:
111.10
Beilstein:
1751158
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

30 mmHg ( 80 °C)

Assay

98%

form

liquid

contains

0.05% 2,6-di-tert-butyl-p-cresol as stabilizer

refractive index

n20/D 1.426 (lit.)

bp

80-82 °C/30 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OC(=C)C#N

InChI

1S/C5H5NO2/c1-4(3-6)8-5(2)7/h1H2,2H3

InChI key

LFSHREXVLSTLFB-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service