Skip to Content
Merck
All Photos(1)

Key Documents

224952

Sigma-Aldrich

1-Piperonylpiperazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$322.00

About This Item

Empirical Formula (Hill Notation):
C12H16N2O2
CAS Number:
Molecular Weight:
220.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$322.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

bp

147-149 °C/2 mmHg (lit.)

mp

36-40 °C (lit.)

SMILES string

C1CN(CCN1)Cc2ccc3OCOc3c2

InChI

1S/C12H16N2O2/c1-2-11-12(16-9-15-11)7-10(1)8-14-5-3-13-4-6-14/h1-2,7,13H,3-6,8-9H2

InChI key

NBOOZXVYXHATOW-UHFFFAOYSA-N

General description

The effect of 1-piperonylpiperazine on 3,4-methylenedioxymethamphetamine (MDMA) induced neurotoxicity was studied[1].

Application

1-Piperonylpiperazine was used in the synthesis of acetyl-caffeic acid-1-piperonylpiperazine (HBU-47)[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seon-Young Park et al.
International immunopharmacology, 19(1), 60-65 (2013-12-24)
In the present study, we synthesized a new hybrid compound by coupling caffeic acid and 1-piperonylpiperazine. The synthetic compound, acetyl-caffeic acid-1-piperonylpiperazine (HBU-47), showed potent anti-inflammatory effects inhibiting lipopolysaccharide (LPS)-induced production of nitric oxide (NO) in RAW264.7 macrophage cells. HBU-47 inhibited
K Hashimoto et al.
European journal of pharmacology, 228(2-3), 171-174 (1992-09-01)
The effects of 1-piperonylpiperazine and N,alpha-dimethylpiperonylamine, which are weak inhibitors for [3H]5-hydroxytryptamine (5-HT) uptake, on 3,4-methylenedioxymethamphetamine (MDMA)-induced neurotoxicity were examined. The reductions of serotonergic parameters in the rat cerebral cortex produced by multiple administration of MDMA (10 mg/kg) were attenuated
Lilian H J Richter et al.
Journal of pharmaceutical and biomedical analysis, 143, 32-42 (2017-06-12)
Metabolism studies play an important role in clinical and forensic toxicology. Because of potential species differences in metabolism, human samples are best suitable for elucidating metabolism. However, in the case of new psychoactive substances (NPS), human samples of controlled studies
K Hashimoto et al.
Brain research, 590(1-2), 341-344 (1992-09-11)
The neurotoxicity of 3,4-methylenedioxymethamphetamine (MDMA) in rat brain was attenuated significantly by coadministration of several benzylpiperazines (p-nitrobenzylpiperazine, p-chlorobenzylpiperazine and 1-piperonylpiperazine), which were weak inhibitors for [3H]6-nitroquipazine binding to the 5-hydroxytryptamine (5-HT) transporter in rat brain. These results suggest that these
Marcelo Dutra Arbo et al.
Archives of toxicology, 90(12), 3045-3060 (2016-01-29)
The piperazine derivatives most frequently consumed for recreational purposes are 1-benzylpiperazine, 1-(3,4-methylenedioxybenzyl) piperazine, 1-(3-trifluoromethylphenyl) piperazine and 1-(4-methoxyphenyl) piperazine. Generally, they are consumed as capsules, tablets or pills but also in powder or liquid forms. Currently, the precise mechanism by which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service