Skip to Content
Merck
All Photos(1)

Key Documents

224049

Sigma-Aldrich

2-Chloro-3,5-dinitropyridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2ClN3O4
CAS Number:
Molecular Weight:
203.54
Beilstein:
196026
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

63-65 °C (lit.)

SMILES string

[O-][N+](=O)c1cnc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C5H2ClN3O4/c6-5-4(9(12)13)1-3(2-7-5)8(10)11/h1-2H

InChI key

QLHVJBXAQWPEDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(1), 147-154 (2005-12-07)
This work deals with the vibrational spectroscopy of 4-amino-2,6-dichloropyridine (ADCP) and 2-chloro-3,5-dinitropyridine (CDNP) by means of quantum chemical calculations. The mid and far FTIR and FT-Raman spectra were measured in the condensed state. The fundamental vibrational frequencies and intensity of
Karl J Okolotowicz et al.
Bioorganic & medicinal chemistry, 23(17), 5282-5292 (2015-08-19)
Cardiomyopathy is the leading cause of death worldwide. Despite progress in medical treatments, heart transplantation is one of the only current options for those with infarcted heart muscle. Stem cell differentiation technology may afford cell-based therapeutics that may lead to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service