Skip to Content
Merck
All Photos(1)

Key Documents

222712

Sigma-Aldrich

2-Fluorocinnamic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$44.10

$44.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$44.10

About This Item

Linear Formula:
FC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
166.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$44.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

178-180 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)\C=C\c1ccccc1F

InChI

1S/C9H7FO2/c10-8-4-2-1-3-7(8)5-6-9(11)12/h1-6H,(H,11,12)/b6-5+

InChI key

IOUDZAFBPDDAMK-AATRIKPKSA-N

Application

2-Fluorocinnamic acid was used in the synthesis of acrylamide[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre L'Heureux et al.
Bioorganic & medicinal chemistry letters, 15(2), 363-366 (2004-12-18)
Replacement of the morpholinyl moiety in (S,E)-N-[1-(3-morpholinophenyl)ethyl]-3-phenylacrylamide (1) with heteroaryl groups led to the identification of (S,E)-N-1-[3-(6-fluoropyridin-3-yl)phenyl]ethyl-3-(2-fluorophenyl)acrylamide (5) as a potent KCNQ2 potassium channel opener. Among this series of heteroaryl substituted acrylamides, (S,E)-N-1-[3-(1H-pyrazol-1-yl)phenyl]ethyl-3-(2-fluorophenyl)acrylamide (9) exhibits balanced potency and efficacy. The
Marina Marinović et al.
Molecules (Basel, Switzerland), 25(19) (2020-09-27)
Harmicines represent hybrid compounds composed of β-carboline alkaloid harmine and cinnamic acid derivatives (CADs). In this paper we report the synthesis of amide-type harmicines and the evaluation of their biological activity. N-harmicines 5a-f and O-harmicines 6a-h were prepared by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service