Skip to Content
Merck
All Photos(1)

Key Documents

222038

Sigma-Aldrich

4-Butylbenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$106.00

$106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$106.00

About This Item

Linear Formula:
CH3(CH2)3C6H4COCl
CAS Number:
Molecular Weight:
196.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$106.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5351 (lit.)

bp

155-156 °C/22 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCCCc1ccc(cc1)C(Cl)=O

InChI

1S/C11H13ClO/c1-2-3-4-9-5-7-10(8-6-9)11(12)13/h5-8H,2-4H2,1H3

InChI key

OUOWCSJYDCPVDM-UHFFFAOYSA-N

Application

4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer)[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pragliola S, et al.
Macromolecular Chemistry and Physics, 200, 2338-2344 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service