Skip to Content
Merck
All Photos(1)

Documents

221058

Sigma-Aldrich

Tris(dimethylamino)methane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3CH
CAS Number:
Molecular Weight:
145.25
Beilstein:
1901162
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

42-43 °C/12 mmHg (lit.)

density

0.838 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)C(N(C)C)N(C)C

InChI

1S/C7H19N3/c1-8(2)7(9(3)4)10(5)6/h7H,1-6H3

InChI key

MUMVIYLVHVCYGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

NMR spectra and their temperature-dependence has been reported for tris(dimethylamino)methane. Tris(dimethylamino)methane reacts with crowded α-(o-nitrophenyl) ketone to yield tetrahydroquinoline derivative.

Application

Tris(dimethylamino)methane was used:
  • to prepare the phosphoramidite derivative of the new nucleoside analog
  • as reagent for the introduction of the dimethylaminoethylidene group
  • in ring-opening (RO) polymerization of lactide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Synthesis & Catalysis, 346, 1081-1081 (2004)
yDNA: a new geometry for size-expanded base pairs.
Haige Lu et al.
Angewandte Chemie (International ed. in English), 43(43), 5834-5836 (2004-11-04)
Conformational Dynamics of Triple Rotors: Tris (dimethylamino) methane, Triisopropylamine, and Related Molecules1.
Anderson JE, et al.
The Journal of Organic Chemistry, 61(4), 1290-1296 (1996)
Sci. Synth., 22, 795-795 (2005)
Tetrahedron, 45, 6631-6631 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service