Skip to Content
Merck
All Photos(1)

Key Documents

220973

Sigma-Aldrich

Titanium(III) chloride

ReagentPlus®, powder

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

product line

ReagentPlus®

form

powder

reaction suitability

reagent type: catalyst
core: titanium

mp

440 °C (dec.) (lit.)

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used for the vapor-phase formation of intermetallic compounds with ultrafine particle size.
Used with tert-butylhydroperoxide in an alcoholic solvent to promote the reductive coupling of aryl aldehydes to 1,2-diols.[1]

Catalyst for:
hydrogen desorption properties[2]
Nitrogen-nitrogen reductive bond cleavage[3]

Reagent for photocatalyst synthesis[4]

Precursor for the preparation of nanostructures titania/cetyltrimethyammonium bromide nanoskeletons[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Organic Chemistry, 4050-4050 (2007)
S. Isobe, et al.,
Applied Physics Letters, 99 (2011)
Shuxi Dai et al.
Nanoscale research letters, 5(11), 1829-1835 (2010-12-03)
Highly crystalline TiO(2) nanostructures were prepared through a facile inorganic acid-assisted hydrothermal treatment of hexagonal-structured assemblies of nanocrystalline titiania templated by cetyltrimethylammonium bromide (Hex-ncTiO(2)/CTAB Nanoskeleton) as starting materials. All samples were characterized by X-ray diffraction (XRD) and transmission electron microscopy
D. Meroni, et al
Catalysis Today, 161, 169-174 (2011)
Gnana S Siluvai et al.
Biochemistry, 48(51), 12133-12144 (2009-11-20)
Sco-like proteins contain copper bound by two cysteines and a histidine residue. Although their function is still incompletely understood, there is a clear involvement with the assembly of cytochrome oxidases that contain the Cu(A) center in subunit 2, possibly mediating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service