Skip to Content
Merck
All Photos(1)

Key Documents

219908

Sigma-Aldrich

4-Imidazolemethanol hydrochloride

98%

Synonym(s):

4-(Hydroxymethyl)imidazole hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$169.00

$169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$169.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2O · HCl
CAS Number:
Molecular Weight:
134.56
Beilstein:
3562041
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$169.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

108-111 °C (lit.)

solubility

water: soluble 50 mg/mL, clear to slightly hazy, colorless to yellow

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

Cl[H].OCc1c[nH]cn1

InChI

1S/C4H6N2O.ClH/c7-2-4-1-5-3-6-4;/h1,3,7H,2H2,(H,5,6);1H

InChI key

WFNASTYGEKUMIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Imidazolemethanol hydrochloride was used in the synthesis of imidazole-containing peptidomimetic inhibitors[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Junko Ohkanda et al.
Organic & biomolecular chemistry, 4(3), 482-492 (2006-02-01)
A series of imidazole-containing peptidomimetic PFTase inhibitors and their co-crystal structures bound to PFTase and FPP are reported. The structures reveal that the peptidomimetics adopt a similar conformation to that of the extended CVIM tetrapeptide, with the imidazole group coordinating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service