All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor pressure
1 mmHg ( 487 °C)
Assay
99.99% trace metals basis
form
mossy
resistivity
5.8 μΩ-cm, 20°C
particle size
1-2 cm
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A viewpoint about the treatment of Wilson's disease.
Abdul Qayyum Rana et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 612-614 (2013-06-22)
Nalini Pandey et al.
Indian journal of experimental biology, 51(7), 548-555 (2013-08-01)
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Michael Alvares et al.
Pediatrics, 132(1), e229-e232 (2013-06-05)
As food allergies become increasingly prevalent and testing methods to identify "food allergy" increase in number, the importance of careful diagnosis has become even more critical. Misdiagnosis of food allergy and inappropriate use of unproven testing modalities may lead to
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service