Skip to Content
Merck
All Photos(1)

Key Documents

214965

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in heptane

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$117.00
800 ML
$425.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$117.00
800 ML
$425.00

About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein:
4123663
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in heptane

density

0.731 g/mL at 25 °C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diisobutylaluminum hydride (DIBAL-H) is a common reducing agent to reduce aldehydes, ketones, esters, acids and acid chlorides to the corresponding alcohols.[1] Some of the applications are:
  • Synthesis of α-acetoxy ethers by reduction and subsequent acetylation of esters.[2]
  • Synthesis of coniferyl, sinapyl, and p-coumaryl alcohol by selective reduction of corresponding cinnamate esters.[3]
  • Reduction of secondary phosphine oxides (SPOs) to the corresponding phosphines.[4]
  • DIBAL-H can also be used in the hydroalumination of alkene and alkynes.[5]

Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of diisobutylaluminium hydride (DIBAH) and triisobutylaluminium (TIBA) as reducing agents in organic synthesis.
Winterfeldt E
Synthesis, 1975(09), 617-630 (1975)
General Synthesis of α-Acetoxy Ethers from Esters by DIBALH Reduction and Acetylation.
Dahanukar V H and Rychnovsky S D
The Journal of Organic Chemistry, 61(23), 8317-8320 (1996)
Facile large-scale synthesis of coniferyl, sinapyl, and p-coumaryl alcohol.
Quideau S and Ralph J.
Journal of Agricultural and Food Chemistry, 40(7), 1108-1110 (1992)
Organometallic compounds of Group III. XIX. Regiospecificity and stereochemistry in the hydralumination of unsymmetrical acetylenes. Controlled cis or trans reduction of 1-alkynyl derivatives.
Eisch J J and Foxton M W
The Journal of Organic Chemistry, 36(23), 3520-3526 (1971)
A Superior method for the reduction of secondary phosphine oxides.
Busacca C A, et al.
Organic Letters, 7(19), 4277-4280 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service