Skip to Content
Merck
All Photos(1)

Key Documents

213810

Sigma-Aldrich

Diethylcyanamide

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$184.00

$184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$184.00

About This Item

Linear Formula:
(C2H5)2NCN
CAS Number:
Molecular Weight:
98.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$184.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

186-188 °C (lit.)

density

0.846 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(CC)C#N

InChI

1S/C5H10N2/c1-3-7(4-2)5-6/h3-4H2,1-2H3

InChI key

ZZTSQZQUWBFTAT-UHFFFAOYSA-N

Gene Information

General description

Diethylcyanamide reacts with hexane solution of lithium diethylamide to yield lithium 1,1,3,3-tetraethylguanidinate[1].

Application

Diethylcyanamide was used in the preparation of N,N′−substituted-5,5′−diamino-3,3′-bi-1,2,4-oxadiazoles[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott D Bunge et al.
Inorganic chemistry, 48(16), 8037-8043 (2009-07-25)
Diethylcyanamide is added to a hexanes solution of lithium diethylamide [LiN(CH(2)CH(3))(2)] resulting in the formation of lithium 1,1,3,3-tetraethylguanidinate, [Li(mu-TEG)](6) (1). Upon successful isolation of 1, the metathesis reaction of MX(2) (MX(2) = MnBr(2), FeBr(2), CoBr(2), and ZnCl(2)) with [Li(mu-TEG)](6) and
N, N′-disubstituted-5, 5′-diamino-3, 3′-bi-1,2,4-oxadiazoles.
Witteveen AJ, et al.
Rec. Trav. Chim., 89(2), 121-128 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service