Skip to Content
Merck
All Photos(4)

Key Documents

211761

Sigma-Aldrich

m-Cresol Purple sodium salt

Dye content 90 %

Synonym(s):

m-Cresolsulfonphthalein sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$74.40
5 G
$141.00
10 G
$295.00

$74.40


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$74.40
5 G
$141.00
10 G
$295.00

About This Item

Empirical Formula (Hill Notation):
C21H17NaO5S
CAS Number:
Molecular Weight:
404.41
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$74.40


Estimated to ship on17 April 2025Details


Request a Bulk Order

form

powder

Quality Level

composition

Dye content, 90%

color

black

λmax

579 nm

ε (extinction coefficient)

≥15000 at 433-439 nm in H2O at 0.008 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1cc(O)ccc1\C(=C2\C=CC(=O)C=C2C)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H18O5S.Na/c1-13-11-15(22)7-9-17(13)21(18-10-8-16(23)12-14(18)2)19-5-3-4-6-20(19)27(24,25)26;/h3-12,22H,1-2H3,(H,24,25,26);/q;+1/p-1/b21-18+;

InChI key

BYQJYOOKENJSNK-GOSREXKOSA-M

Looking for similar products? Visit Product Comparison Guide

General description

m-cresol purple sodium salt (MCP) is a colorimetric pH indicator dye. The dye is blue in anionic form and yellow in protonated form.[1]

Application

m-Cresol Purple sodium salt has been used to measure pH of seawater using a spectrophotometer.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric high-precision seawater pH determination
for use in underway measuring systems
A?mann S
Ocean Science, 7, 597-607 (2011)
M.-I. Baraton
Sensors for Environment, Health and Security (2008)
Peter J Edmunds et al.
The Journal of experimental biology, 223(Pt 8) (2020-03-18)
Experiments with coral fragments (i.e. nubbins) have shown that net calcification is depressed by elevated PCO2 Evaluating the implications of this finding requires scaling of results from nubbins to colonies, yet the experiments to codify this process have not been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service