Skip to Content
Merck
All Photos(1)

Key Documents

21155

Sigma-Aldrich

Calcium α-D-heptagluconate hydrate

≥98.0% (calc. based on dry substance)

Synonym(s):

α-D-Glucoheptonic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26CaO16 · xH2O
CAS Number:
Molecular Weight:
490.42 (anhydrous basis)
Beilstein:
6104529
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (calc. based on dry substance)

form

powder

optical activity

[α]20/D +2±0.2°, c = 5% in H2O

loss

~5% loss on drying

SMILES string

O.O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C7H14O8.Ca.2H2O/c2*8-1-2(9)3(10)4(11)5(12)6(13)7(14)15;;;/h2*2-6,8-13H,1H2,(H,14,15);;2*1H2/q;;+2;;/p-2/t2*2-,3-,4+,5-,6-;;;/m11.../s1

InChI key

JBMIEBJMNFOKBC-JRLDONDGSA-L

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Gale et al.
Nuclear medicine communications, 13(2), 110-113 (1992-02-01)
This study attempted to evaluate the role of glucoheptonate (GHA) in captopril renography in an in vivo laboratory investigation in which postcaptopril glucoheptonate uptake was analysed in awake 2KlC hypertensive rats. Clamped kidney uptake in a previous study was greater
[Comparison of the bioavailability of fluorine from sodium fluoride alone and in combination with various calcium salts].
D Briançon et al.
Revue du rhumatisme et des maladies osteo-articulaires, 56(1), 83-86 (1989-01-01)
M K Gumerlock et al.
Neurosurgery, 26(2), 268-277 (1990-02-01)
To evaluate the effect of various anesthetic agents on hyperosmolar blood-brain barrier disruption (BBBD), Sprague-Dawley rats were given pentobarbital (PB), ketamine-xylazine (KX), isoflurane (IF), methoxyflurane (MF), or fentanyl-droperidol (FD) before intracarotid infusion of mannitol or saline. Physiological monitoring showed that
I M McPhee et al.
The Veterinary record, 122(20), 483-485 (1988-05-14)
Trials were carried out to compare the efficacy of copper heptonate with other available therapeutic products. On nine farms with a copper deficiency problem, over 2000 ewes were treated in mid-pregnancy. All the treatments increased whole blood copper levels and
Glucoheptonate for diagnosis of brain tumor.
V Wiwanitkit
Journal of postgraduate medicine, 56(2), 168-168 (2010-07-14)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service