Skip to Content
Merck
All Photos(1)

Key Documents

21057

Sigma-Aldrich

Calcium borohydride bis-tetrahydrofuran complex

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(BH4)2 · 2C4H8O
CAS Number:
Molecular Weight:
213.97
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥98.0%

reaction suitability

reagent type: reductant

SMILES string

[BH4-].[BH4-].[Ca++].C1CCOC1.C2CCOC2

Other Notes

Ready-to-use, selective reducing agent[1][2]; e.g. mild reduction of esters[3][4]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Ogawa et al.
Synthesis, 459-459 (1990)
G.G. Harbermehl et al.
Heterocycles, 7, 1027-1027 (1977)
J.-P. Barnier et al.
Tetrahedron, 45, 5051-5051 (1989)
M Fujita et al.
Journal of medicinal chemistry, 33(7), 1898-1905 (1990-07-01)
As an extension of the previous investigation (J. Med. Chem. 1988, 31, 919), we synthesized a series of 2-[2-[(aminoalkyl)oxy]-5-methoxyphenyl]-3,4-dihydro-4-methyl-3-oxo-2H- 1,4-benzothiazines (3) and evaluated their Ca2+ antagonistic activities. Ca2+ antagonistic activity was measured with isolated depolarized guinea pig taenia cecum. On

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service