Skip to Content
Merck
All Photos(1)

Documents

210218

Sigma-Aldrich

3-(Dimethylamino)-1,2-propanediol

98%

Synonym(s):

(±)-3-(Dimethylamino)-1,2-propanediol, 1,2-Dihydroxy-3-(dimethylamino)propane, Methicol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
119.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4609 (lit.)

bp

216-217 °C (lit.)

density

1.004 g/mL at 25 °C (lit.)

SMILES string

CN(C)CC(O)CO

InChI

1S/C5H13NO2/c1-6(2)3-5(8)4-7/h5,7-8H,3-4H2,1-2H3

InChI key

QCMHUGYTOGXZIW-UHFFFAOYSA-N

Related Categories

Application

3-(Dimethylamino)-1,2-propanediol was used in the preparation of:
  • synthetic cationic lipid, N-[1-(2,3-dioleyloxy)propyl-N,N,N-trimethylammonium chloride (DOTMA), used in DNA-transfection protocol
  • indane-derived bis(oxazolines)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P L Felgner et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(21), 7413-7417 (1987-11-01)
A DNA-transfection protocol has been developed that makes use of a synthetic cationic lipid, N-[1-(2,3-dioleyloxy)propyl]-N,N,N-trimethylammonium chloride (DOTMA). Small unilamellar liposomes containing DOTMA interact spontaneously with DNA to form lipid-DNA complexes with 100% entrapment of the DNA, DOTMA facilitates fusion of
An efficient synthesis of indane-derived bis (oxazoline) and its application to hetero Diels-Alder reactions on polymer support.
Kurosu M, et al.
Tetrahedron Letters, 45(1), 145-148 (2004)
Shuo Fang et al.
International journal of pharmaceutics, 493(1-2), 460-465 (2015-08-11)
A novel dual drug-tailed betaine conjugate amphiphile has been firstly synthesized in which the polar headgroup is derived from glycine betaine and the hydrophobic tails are chlorambucil molecules. The newly prepared conjugate undergoes self-assembly to form stable liposome-like nanocapsules as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service