Skip to Content
Merck
All Photos(1)

Key Documents

209732

Sigma-Aldrich

3-Phenylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N
CAS Number:
Molecular Weight:
155.20
Beilstein:
110400
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.616 (lit.)

bp

269-270 °C/749 mmHg (lit.)

density

1.082 g/mL at 25 °C (lit.)

SMILES string

c1ccc(cc1)-c2cccnc2

InChI

1S/C11H9N/c1-2-5-10(6-3-1)11-7-4-8-12-9-11/h1-9H

InChI key

HJKGBRPNSJADMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Phenylpyridine forms complexes with gold(III), palladium(II) and platinum(II) chloride and their 1H, 13C and 15N nuclear magnetic resonance studies have been reported[1]. Interactions of 3-phenylpyridine with copper surface has been investigated by Surface-enhanced Raman scattering (SERS) spectroscopy and cyclic voltammetry[2]. It is a useful synthetic intermediate.

Application

3-Phenylpyridine was employed as axial ligand to investigate the rate of olefin oxygenation catalyzed by synthetic metalloporphyrins[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc P Bonaca et al.
Future cardiology, 5(5), 435-442 (2009-09-01)
SCH 530348, a synthetic tricyclic 3-phenylpyridine, is an orally active fourth generation himbacine-based antagonist of the protease-activated receptor (PAR)-1, the primary receptor for thrombin on platelets in humans. SCH 530348 is the first in a new class of compounds that
T L Perry et al.
Journal of the neurological sciences, 85(3), 309-317 (1988-07-01)
Idiopathic Parkinson's disease (PD) is likely to be caused by one or more unidentified neurotoxins, present in the environment or formed endogenously, which progressively damage dopaminergic nigrostriatal neurons. 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) is an experimental neurotoxin which produces biochemical and neuropathological changes
Sanjeev Kumar et al.
Journal of medicinal chemistry, 51(16), 4968-4977 (2008-07-31)
Indoleamine 2,3-dioxygenase (IDO) is emerging as an important new therapeutic target for the treatment of cancer, chronic viral infections, and other diseases characterized by pathological immune suppression. With the goal of developing more potent IDO inhibitors, a systematic study of
Surface-enhanced Raman spectroscopy studies of phenylpyridines interacting with a copper electrode surface.
Zawada K and Bukowska J.
Surface Science, 507, 34-39 (2002)
J P Collman et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(22), 7039-7041 (1983-11-01)
The rate of olefin oxygenation catalyzed by synthetic metalloporphyrins is examined, employing sodium hypochlorite as the oxygen atom source. The rate of epoxidation and the stability of the catalyst are shown to be dependent on the nature of the axial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service