Skip to Content
Merck
All Photos(2)

Key Documents

209066

Sigma-Aldrich

Osmium(III) chloride hydrate

Synonym(s):

Osmium trichloride hydrate, Osmium(3+) trichloride hydrate, Trichloroosmium hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$421.00
2.5 G
$1,650.00

$421.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$421.00
2.5 G
$1,650.00

About This Item

Linear Formula:
OsCl3 · xH2O
CAS Number:
Molecular Weight:
296.59 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$421.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reagent type: catalyst
core: osmium

SMILES string

O.Cl[Os](Cl)Cl

InChI

1S/3ClH.H2O.Os/h3*1H;1H2;/q;;;;+3/p-3

InChI key

KHHOLOMEVBPRQY-UHFFFAOYSA-K

Application

Reagent for preparation of dichlorodihydridoosmium complexes, useful starting materials and catalysts.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aracama, M.
Inorganic Chemistry, 30, 288-288 (1991)
Christopher S Wilson et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(50), 11593-11603 (2020-06-11)
Dinuclear metallodrugs offer much potential in the development of novel anticancer chemotherapeutics as a result of the distinct interactions possible with bio-macromolecular targets and the unique biological activity that can result. Herein, we describe the development of isostructural homo-dinuclear OsII

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service