Skip to Content
Merck
All Photos(1)

Key Documents

205184

Sigma-Aldrich

Hexachlorodisilane

96%

Synonym(s):

1,1,1,2,2,2-Hexachlorodisilane, Disilicon hexachloride, HCDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(SiCl3)2
CAS Number:
Molecular Weight:
268.89
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

144-145.5 °C (lit.)

density

1.562 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)[Si](Cl)(Cl)Cl

InChI

1S/Cl6Si2/c1-7(2,3)8(4,5)6

InChI key

LXEXBJXDGVGRAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexachlorodisilane (HCDS) is a chlorosilane used as a precursor for producing disilanes. It is a dioxidizer that is used in the production of silicon films and silicon nitride based films.[1]

Application

HCDS can be used in the fabrication of silica aerogels by chemical vapor deposition (CVD), which can be potentially used as encapsulating agents and thermal insulators.[2][3] It can also be used to synthesize 1,1,1,2,2,2-hexaamino-disilanes using CVD, which forms silicon-based films for microelectronic-based applications.[4]
HCDS may be used as a reducing agent. It may be combined with ammonia to form silicon nitride by chemical vapor deposition(CVD) technique.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Film Properties of Low?k Silicon Nitride Films Formed by Hexachlorodisilane and Ammonia.
Tanaka M , et al.
Journal of the Electrochemical Society, 147(6), 2284-2289 (2000)
Chemical vapor deposition of silicon films using hexachlorodisilane
Taylor, R. C., Scott, B. A., Lin, S. T., LeGoues, F., & Tsang, J. C.
MRS Proceedings, 77, 709-709 (1986)
Enhancing mechanical properties of silica aerogels
Obrey, K. A., Wilson, K. V., & Loy, D. A.
Journal of Non-Crystalline Solids, 375(19), 3435-3441 (2011)
Enhancing mechanical properties of silica aerogels.
Obrey KAD, et al.
Journal of Non-Crystalline Solids, 357(19-20), 3435-3441 (2011)
Initial Reaction of Hexachlorodisilane on Amorphous Silica Surface for Atomic Layer Deposition Using Density Functional Theory.
Kim K, et al.
Journal of the Korean Ceramic Society, 54(5), 443-447 (2017)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service