Skip to Content
Merck
All Photos(2)

Key Documents

205141

Sigma-Aldrich

Praseodymium(III) chloride hydrate

99.9% trace metals basis

Synonym(s):

Praseodymium Trichloride Hydrate, Praseodymium chloride hydrate, Praseodymium(3+) Trichloride Hydrate, Trichloropraseodymium hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$131.00
50 G
$436.00

$131.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$131.00
50 G
$436.00

About This Item

Linear Formula:
PrCl3 · xH2O
CAS Number:
Molecular Weight:
247.27 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$131.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: praseodymium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

density

2.25 g/mL at 25 °C (lit.)

SMILES string

O.Cl[Pr](Cl)Cl

InChI

1S/3ClH.H2O.Pr/h3*1H;1H2;/q;;;;+3/p-3

InChI key

BQRCNEKQNZWNCY-UHFFFAOYSA-K

General description

Praseodymium (III) chloride hydrate is soluble in water and alcohol. It is used as a source of Pr to prepare insoluble praseodymium (III)compounds.

Application

Praseodymium(III) chloride hydrate can be used as a dopant to prepare Pr-modified TiO2 nanoparticles for photocatalytic degradation of organic pollutants. The addition of Pr ion enhances the thermal stability and surface textural properties of the catalyst.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic activity of Pr-modified TiO2 for the degradation of bisphenol A
Denise S Cordeiro, et al.
SN Applied Sciences, 3, 1-8 (2021)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service