Skip to Content
Merck
All Photos(2)

Key Documents

204870

Sigma-Aldrich

Ytterbium(III) chloride hexahydrate

99.998% trace metals basis

Synonym(s):

Ytterbium trichloride hydrate, Ytterbium(III) chloride·6hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$452.00
50 G
$1,600.00

$452.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$452.00
50 G
$1,600.00

About This Item

Linear Formula:
YbCl3 · 6H2O
CAS Number:
Molecular Weight:
387.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

$452.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

99.998% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤25.0 ppm Trace Rare Earth Analysis

mp

180 °C (dec.) (lit.)

density

2.58 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Yb](Cl)Cl

InChI

1S/3ClH.6H2O.Yb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

LEYFXTUKPKKWMP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ytterbium(III) chloride hexahydrate is a white crystalline solid used as a precursor to synthesize nanomaterials and as acatalyst in various organic transformations.

Application

Ytterbium(III) chloride hexahydrate can be used as a precursor:
  • To synthesize ytterbium oxide nanodiscs to fabricate white LEDs.[1]
  • To prepare NaYF4:Yb/Ho/Mn upconversion nanoparticles with good luminescent intensity, for biomedical applications.[2]
  • To fabricate three-dimensional NaYF4:Yb3+/Tm3+hexagonal prisms for near IR photocatalytic H2 evolution.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of Luminescence Intensity in Red Emitting NaYF4:Yb/Ho/Mn Upconversion Nanophosphors by Variation of Reaction Parameters
Kumbam Lingeshwar Reddy, et al.
The Journal of Physical Chemistry C, 121, 11783-11793 (2017)
Simple one pot synthesis of luminescent europium doped yttrium oxide Y2O3: Eu nanodiscs for phosphor converted warm white LEDs
Julian Petry, et al.
Nanoscale advances, 4, 858-864 (2022)
Three-dimensional NaYF4:Yb3+/Tm3+ hexagonal prisms coupled with ZnIn2S4 nanosheets for boosting near-infrared photocatalytic H2 evolution
Zhouze Chen, et al.
Journal of Environmental Chemical Engineering, 10, 108352-108352 (2022)
Uliana Kostiv et al.
Beilstein journal of nanotechnology, 6, 2290-2299 (2016-01-07)
NaYF4:Yb(3+)/Er(3+) nanoparticles were synthesized by thermal decomposition of lanthanide trifluoroacetates using oleylamine (OM) as both solvent and surface binding ligand. The effect of reaction temperature and time on the properties of the particles was investigated. The nanoparticles were characterized by

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service