Skip to Content
Merck
All Photos(2)

Key Documents

20444

Sigma-Aldrich

(±)-3-tert-Butoxy-1,2-propanediol

≥97.0% (GC)

Synonym(s):

(±)-1-tert-Butylglycerol, (±)-Glycerol 1-tert-butyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$230.00

$230.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$230.00

About This Item

Linear Formula:
(CH3)3COCH2CHOHCH2OH
CAS Number:
Molecular Weight:
148.20
Beilstein:
1698535
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$230.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥97.0% (GC)

refractive index

n20/D 1.44

density

1.0 g/mL at 20 °C (lit.)

functional group

ether
hydroxyl

SMILES string

CC(C)(C)OCC(O)CO

InChI

1S/C7H16O3/c1-7(2,3)10-5-6(9)4-8/h6,8-9H,4-5H2,1-3H3

InChI key

JPWDLYMEUNBLIR-UHFFFAOYSA-N

Application

(±)-3-tert-Butoxy-1,2-propanediol may be used in chemical synthesis studies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service