Skip to Content
Merck
All Photos(2)

Key Documents

204331

Sigma-Aldrich

Ammonium hexafluorosilicate

99.999% trace metals basis

Synonym(s):

Cryptohalite

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 229.07
100 MG
CHF 343.56
250 MG
CHF 521.94

Vendor SKU: W11413-50MG

CHF 229.07


Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
50 MG
CHF 229.07
100 MG
CHF 343.56
250 MG
CHF 521.94

About This Item

Linear Formula:
(NH4)2SiF6
CAS Number:
Molecular Weight:
178.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Vendor SKU: W11413-50MG

CHF 229.07


Ships in 14 days - from Aldrich Partner.

Request a Bulk Order

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

density

2.01 g/mL at 25 °C (lit.)

SMILES string

[H][N+]([H])([H])[H].[H][N+]([H])([H])[H].F[Si--](F)(F)(F)(F)F

InChI

1S/F6Si.2H3N/c1-7(2,3,4,5)6;;/h;2*1H3/q-2;;/p+2

InChI key

ITHIMUMYFVCXSL-UHFFFAOYSA-P

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toshiyuki Suge et al.
Dental materials : official publication of the Academy of Dental Materials, 24(2), 192-198 (2007-06-05)
Diamine silver fluoride [AgF: (NH(3))(2)AgF] has been used clinically in Japan, as it reduces dental caries and dentin hypersensitivity. However, AgF stains the teeth black due to silver precipitation. To overcome this drawback, the authors prepared ammonium hexafluorosilicate [SiF: (NH(4))(2)SiF(6)]
[Recurrent ventricular fibrillation in magnesium fluosilicate poisoning].
B Suberviola Cañas et al.
Medicina intensiva, 31(6), 344-346 (2007-08-01)
Mark A Brzezinski et al.
Analytical chemistry, 78(17), 6109-6114 (2006-09-02)
A procedure for the automated determination of isotopic abundances of silicon from biogenic and lithogenic particulate matter and from dissolved silicon in fresh or saltwaters is reported. Samples are purified using proven procedures through the reaction of Si with acidified
Toshiyuki Suge et al.
American journal of dentistry, 19(4), 248-252 (2006-08-31)
To evaluate the occluding ability of ammonium hexafluorosilicate (SiF). Dentin disks prepared from human extracted teeth were grouped as follows to prepare different situations of dentin hypersensitivity: (1) those sonicated for 20 minutes; (2) those treated with 0.5 mol/L EDTA
Teresa Lech
Forensic science international, 206(1-3), e20-e24 (2010-07-27)
Fluoride, of all inorganic substances, is among the least likely to be identified by a routine toxicological analysis. Acute poisonings with salts of hydrofluoric or fluorosilicic acid, however, although relatively uncommon, may occur. Some fluorosilicates, salts of fluorosilicic acid (e.g.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service