Skip to Content
Merck
All Photos(2)

Key Documents

199893

Sigma-Aldrich

Ethyl 4-methyl-5-imidazolecarboxylate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$282.00

$282.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$282.00

About This Item

Empirical Formula (Hill Notation):
C7H10N2O2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$282.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

204-206 °C (lit.)

solubility

chloroform/methanol: soluble 50 mg/mL, clear, colorless to light yellow

functional group

ester

SMILES string

CCOC(=O)c1[nH]cnc1C

InChI

1S/C7H10N2O2/c1-3-11-7(10)6-5(2)8-4-9-6/h4H,3H2,1-2H3,(H,8,9)

InChI key

VLDUBDZWWNLZCU-UHFFFAOYSA-N

General description

Ethyl 4-methyl-5-imidazolecarboxylate forms coordination compounds with Co(2+). These compounds inhibit photosynthetic electron flow and ATP-synthesis and acts as Hill reaction inhibitors[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz King-Díaz et al.
Bioinorganic chemistry and applications, 93-108 (2008-03-28)
In this work we present the synthesis, structural and spectroscopic characterization of Co(2+) coordination compounds with ethyl 4-methyl-5-imidazolecarboxylate (emizco). The effects of emizco, the metal salts CoCl(2).6H(2)O, CoBr(2), Co(NO(3))(2).6H(2)O and their metal coordination compounds [Co(emizco)(2)Cl(2)], [Co(emizco)(2) Br(2)].H(2)O, [Co(emizco)(2) (H(2)O)(2)(NO(2))(2).2H(2)O were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service