Skip to Content
Merck
All Photos(1)

Key Documents

199400

Sigma-Aldrich

2,3-Dimethylpyrazine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$67.90

$67.90


Available to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$67.90

About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$67.90


Available to ship on24 April 2025Details


Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.507 (lit.)

bp

156 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

SMILES string

Cc1nccnc1C

InChI

1S/C6H8N2/c1-5-6(2)8-4-3-7-5/h3-4H,1-2H3

InChI key

OXQOBQJCDNLAPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The bacterium strain DP-45, identified as Rhodococcus erythropolis, was grown on 2,3-dimethylpyrazine[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sugima Rappert et al.
Biodegradation, 18(5), 585-596 (2006-11-23)
A bacterium, strain DP-45, capable of degrading 2,5-dimethylpyrazine (2,5-DMP) was isolated and identified as Rhodococcus erythropolis. The strain also grew on many other pyrazines found in the waste gases of food industries, like 2,3-dimethylpyrazine (2,3-DMP), 2,6-dimethylpyrazine (2,6-DMP), 2-ethyl-5(6)-dimethylpyrazine (EMP), 2-ethylpyrazine
Mario Silva et al.
Journal of chromatography. A, 1629, 461508-461508 (2020-08-29)
In the present document, we report the development of an analytical method consisting of a sequential direct-immersion/headspace solid-phase microextraction (DI-HS-SPME) followed by gas-phase chromatography and tandem mass spectrometry (GC-MS/MS) for simultaneous analysis of 4-chlorobenzyl alcohol, 2,6-dichlorobenzyl alcohol, 4-methoxybenzyl alcohol, 3,4-dimethoxybenzyl
Nobuo Ochiai et al.
Journal of chromatography. A, 1421, 103-113 (2015-08-16)
An extension of multi-volatile method (MVM) technology using the combination of a standard dynamic headspace (DHS) configuration, and a modified DHS configuration incorporating an additional vacuum module, was developed for milliliter injection volume of aqueous sample with full sample evaporation.
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service