Skip to Content
Merck
All Photos(1)

Key Documents

198188

Sigma-Aldrich

1,5-Hexanediol

99%

Synonym(s):

1,5-Dihydroxyhexane, 1-Methyl-1,5-pentanediol, Hexan-1,5-diol, Hexane-2,6-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$146.00
5 G
$503.00

$146.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$146.00
5 G
$503.00

About This Item

Linear Formula:
CH3CH(OH)(CH2)4OH
CAS Number:
Molecular Weight:
118.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$146.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.451 (lit.)

bp

89-91 °C/0.5 mmHg (lit.)

density

0.981 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)CCCCO

InChI

1S/C6H14O2/c1-6(8)4-2-3-5-7/h6-8H,2-5H2,1H3

InChI key

UNVGBIALRHLALK-UHFFFAOYSA-N

General description

Surface tension and binding constants were studied for 1,5-hexanediol[1].

Application

  • Metadynamics of polyalcohol dehydration in water: This study provides insights into the dehydration mechanisms of polyalcohols like 1,5-Hexanediol, relevant for understanding their behavior in aqueous environments, useful for drug delivery systems and biomedical applications (Ruiz-Barragan et al., 2015).
  • Direct Copolymerization of CO2 and Diols: This research explores the copolymerization of CO2 with diols including 1,5-Hexanediol, pointing to sustainable polymer production methods that are crucial for material science (Tamura et al., 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of temperature on the surface tension of diluted aqueous solutions of 1, 2-hexanediol, 1, 5-hexanediol, 1, 6-hexanediol and 2, 5-hexanediol.
Fluid Phase Equilibria, 258(1), 67-72 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service