Skip to Content
Merck
All Photos(1)

Key Documents

197556

Sigma-Aldrich

4-(3-Methyl-5-oxo-2-pyrazolin-1-yl)benzoic acid

≥98%

Synonym(s):

1-(4-Carboxyphenyl)-3-methyl-2-pyrazolin-5-one, 1-(4-Carboxyphenyl)-3-methyl-2-pyrazolinone, 1-(4-Carboxyphenyl)-3-methyl-5-pyrazolone, 1-(p-Carboxyphenyl)-3-methyl-5-pyrazolone, 4-(3-Methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid, 4-(3-Methyl-5-oxopyrazol-1-yl)benzoic acid, 4-(4,5-Dihydro-3-methyl-5-oxo-1H-pyrazol-1-yl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$100.00

$100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$100.00

About This Item

Empirical Formula (Hill Notation):
C11H10N2O3
CAS Number:
Molecular Weight:
218.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

mp

285 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

CC1=NN(C(=O)C1)c2ccc(cc2)C(O)=O

InChI

1S/C11H10N2O3/c1-7-6-10(14)13(12-7)9-4-2-8(3-5-9)11(15)16/h2-5H,6H2,1H3,(H,15,16)

InChI key

CUGBBQWDGCXWNB-UHFFFAOYSA-N

General description

The pharmacological activity of 4-(3-methyl-5-oxo-2-pyrazolin-1-yl)benzoic acid was studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Antón-Fos et al.
Arzneimittel-Forschung, 44(7), 821-826 (1994-07-01)
New compounds showing hypoglycaemic activity have been designed through a computer-aided method based on QSAR (quantitative structure activity relationship) and molecular connectivity. The pharmacological tests carried out on the newly designed compounds demonstrated the existence of notable activity for two
Xixi Chen et al.
EBioMedicine, 25, 143-153 (2017-10-17)
Glycated haemoglobin (HbA1c) is the most important marker of hyperglycaemia in diabetes mellitus. We show that d-ribose reacts with haemoglobin, thus yielding HbA1c. Using mass spectrometry, we detected glycation of haemoglobin with d-ribose produces 10 carboxylmethyllysines (CMLs). The first-order rate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service