Skip to Content
Merck
All Photos(2)

Key Documents

197181

Sigma-Aldrich

2,5-Dimethoxy-4-nitroaniline

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$108.00

$108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$108.00

About This Item

Linear Formula:
O2NC6H2(OCH3)2NH2
CAS Number:
Molecular Weight:
198.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$108.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

mp

152-155 °C (lit.)

functional group

nitro

SMILES string

COc1cc(c(OC)cc1N)[N+]([O-])=O

InChI

1S/C8H10N2O4/c1-13-7-4-6(10(11)12)8(14-2)3-5(7)9/h3-4H,9H2,1-2H3

InChI key

ZTQGTYFYOODGOQ-UHFFFAOYSA-N

General description

2,5-Dimethoxy-4-nitroaniline was used in screening of enzyme arylamine N-acetyltransferase isolated from Bacillus cereus[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Hasmann et al.
Archives of microbiology, 146(3), 275-279 (1986-12-01)
The raw extracts of a series of microorganisms were screened for the presence of acetyl-coenzyme A: arylamine N-acetyltransferase (AAAT) using a radioactive assay with 3H-acetyl-coenzyme A and aniline as substrates. Enzyme activities were primarily detected in the soluble fractions of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service