Skip to Content
Merck
All Photos(1)

Key Documents

197173

Sigma-Aldrich

2-Hydrazino-2-imidazoline hydrobromide

98%

Synonym(s):

2-Hydrazino-4,5-dihydroimidazole hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$286.00

$286.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$286.00

About This Item

Empirical Formula (Hill Notation):
C3H8N4 · HBr
CAS Number:
Molecular Weight:
181.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$286.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

98%

mp

185-187 °C (lit.)

SMILES string

Br.NNC1=NCCN1

InChI

1S/C3H8N4.BrH/c4-7-3-5-1-2-6-3;/h1-2,4H2,(H2,5,6,7);1H

InChI key

HIJRJVVSABWXKG-UHFFFAOYSA-N

Application

2-Hydrazino-2-imidazoline hydrobromide was used in synthesis of:
  • some amide derivatives of 6-(4-carboxymethyloxyphenyl)-4,5-dihydro-3(2H)-pyridazinone[1]
  • 2-substituted-6-(4-acylaminophenyl)-4,5-dihydropyridazin-3(2H)-ones, potential inodilating agents[2]
Reactant for synthesis of:
  • Molecules that affect vasodilatory activity
  • Sulfate-bridges dimerica copper(II) complexes
  • 5-HT6 serotonin receptor antagonists
  • Pd complexes
  • Antidiabetic/antiobesity agents
  • DNA binders

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranju Bansal et al.
European journal of medicinal chemistry, 44(11), 4441-4447 (2009-07-11)
Synthesis and vasodilatory activity of some amide derivatives of 6-(4-carboxymethyloxyphenyl)-4,5-dihydro-3(2H)-pyridazinone are reported. An effect of substitution at 2-position of pyridazinone ring on vasodilatory potential has also been explored. The most active compound 6-[4-(2-oxo-2-pyrrolidin-1-yl-ethoxy)phenyl]-2-(4-fluorophenyl)-4,5-dihydropyridazin-3(2H)-one (11) exhibited vasodilating activity in nanomolar range
Dinesh Kumar et al.
Acta pharmaceutica (Zagreb, Croatia), 58(4), 393-405 (2008-12-24)
The present study describes the synthesis and pharmacological evaluation of 2-substituted-6-(4-acylaminophenyl)-4,5-dihydropyridazin-3(2H)-ones as potent inodilating agents. The synthesis of target compounds 2-4 and 7-11 was achieved by Friedel-Crafts acylation of appropriate anilide derivative with succinic anhydride or methylsuccinic anhydride and subsequent
Mohamed A Hashem et al.
Carbohydrate polymers, 237, 116177-116177 (2020-04-04)
A new hydrazono-imidazoline modified cellulose (HIMC) was synthesized for selective recovery of Pt(IV), Pd(II) and Au(III) from geological samples. Cellulose was oxidized by periodate and was further functionalized with hydrazono-imidazoline moieties to afford N-donor chelating fibers. Scanning electron microscopy (SEM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service