Skip to Content
Merck
All Photos(2)

Key Documents

195898

Sigma-Aldrich

3-(4-Methoxyphenoxy)benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OC6H4CHO
CAS Number:
Molecular Weight:
228.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.596 (lit.)

bp

145 °C/0.4 mmHg (lit.)

density

1.089 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

COc1ccc(Oc2cccc(C=O)c2)cc1

InChI

1S/C14H12O3/c1-16-12-5-7-13(8-6-12)17-14-4-2-3-11(9-14)10-15/h2-10H,1H3

InChI key

WLFDEVVCXPTAQA-UHFFFAOYSA-N

Application

3-(4-Methoxyphenoxy)benzaldehyde was used as building block in the synthesis of tetrahydroisoquinolinones[1]. It was used as internal standard during the determination of hydrolytic activity of pyrethroids by gas chromatography-mass spectrometry[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Nishi et al.
Archives of biochemistry and biophysics, 445(1), 115-123 (2005-12-20)
Carboxylesterases hydrolyze a large array of endogenous and exogenous ester-containing compounds, including pyrethroid insecticides. Herein, we report the specific activities and kinetic parameters of human carboxylesterase (hCE)-1 and hCE-2 using authentic pyrethroids and pyrethroid-like, fluorescent surrogates. Both hCE-1 and hCE-2
M Lebl
Bioorganic & medicinal chemistry letters, 9(9), 1305-1310 (1999-05-26)
A new technique for high throughput solid phase synthesis using the centrifuge based liquid removal from readily available standard microtiterplates is described. This technique eliminates the filtration step and is therefore applicable to simultaneous processing of an unlimited number of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service