Skip to Content
Merck
All Photos(3)

Key Documents

194972

Sigma-Aldrich

tert-Butylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$38.50
100 G
$131.00
500 G
$231.00

$38.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$38.50
100 G
$131.00
500 G
$231.00

About This Item

Linear Formula:
(CH3)3CNHNH2 · HCl
CAS Number:
Molecular Weight:
124.61
Beilstein:
3651224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$38.50


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

191-194 °C (lit.)

functional group

amine
hydrazine

SMILES string

Cl.CC(C)(C)NN

InChI

1S/C4H12N2.ClH/c1-4(2,3)6-5;/h6H,5H2,1-3H3;1H

InChI key

DDPWVABNMBRBFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butylhydrazine hydrochloride was used in the synthesis of:
  • diphosphinohydrazines[1]
  • 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole[2]
  • 1,3,4,5-tetrasubstituted pyrazole derivatives[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective synthesis of 1, 3, 4, 5-tetrasubstituted pyrazoles from Baylis-Hillman adducts.
Lee KY, et al.
Tetrahedron Letters, 44(35), 6737-6740 (2003)
One-pot synthesis of tetrasubstituted pyrazoles-Proof of regiochemistry.
Hanefeld U, et al.
Journal of the Chemical Society. Perkin Transactions 1, 13, 1545-1552 (1996)
Alexander N Kornev et al.
Inorganic chemistry, 51(2), 874-881 (2012-01-03)
Reactions of diphosphinohydrazines R-NH-N(PPh(2))(2) (R = tBu (1), Ph(2)P (3)) with some metalation reagents (Co[N(SiMe(3))(2)](2), LiN(SiMe(3))(2), La[N(SiMe(3))(2)](3), nBuLi, MeLi) were performed. Compound 1 was synthesized by the reaction of Ph(2)PCl with tert-butylhydrazine hydrochloride in 83% yield. This compound reveals temperature-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service