Skip to Content
Merck
All Photos(1)

Key Documents

194824

Sigma-Aldrich

Borane dimethyl sulfide complex solution

2.0 M in toluene

Synonym(s):

BMS, Trihydro[thiobis[methane]]boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Beilstein:
3663489
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

reaction suitability

reagent type: reductant

concentration

2.0 M in toluene

density

0.856 g/mL at 25 °C

SMILES string

B.CSC

InChI

1S/C2H6S.BH3/c1-3-2;/h1-2H3;1H3

InChI key

RMHDLBZYPISZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant used as a regioselective reducing agent

Reactant involved in:
  • Hydroboration / oxidation
Synthesis of cage compounds

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen C Rollet-Kurhajec et al.
PloS one, 10(6), e0129868-e0129868 (2015-06-20)
In Hepatitis C virus (HCV) mono-infection, male sex is associated with faster liver fibrosis progression but the effects of sex have not been well studied in HIV-HCV co-infected patients. We examined the influence of sex on progression to significant liver
A Burchert et al.
Leukemia, 29(6), 1331-1335 (2015-02-26)
A minority of chronic myeloid leukemia (CML) patients is capable of successfully discontinuing imatinib. Treatment modalities to increase this proportion are currently unknown. Here, we assessed the role of interferon alpha 2a (IFN) on therapy discontinuation in a previously reported
Andrew J Saykin et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 11(7), 792-814 (2015-07-22)
Genetic data from the Alzheimer's Disease Neuroimaging Initiative (ADNI) have been crucial in advancing the understanding of Alzheimer's disease (AD) pathophysiology. Here, we provide an update on sample collection, scientific progress and opportunities, conceptual issues, and future plans. Lymphoblastoid cell
Saskia Ditisheim et al.
Inflammatory bowel diseases, 21(11), 2598-2604 (2015-08-06)
Inflammatory bowel diseases (IBD) are systemic conditions that commonly display extraintestinal manifestations. Inflammatory articular disease (IAD: axial or peripheral) is the most common extraintestinal manifestation. The aim of this study was to evaluate the prevalence and the clinical characteristics associated
Veena K Ranganath et al.
Arthritis care & research, 67(7), 929-939 (2015-01-13)
To evaluate the effect of sustained American College of Rheumatology (ACR)/European League Against Rheumatism (EULAR) Boolean remission on residual joint inflammation assessed by magnetic resonance imaging (MRI) and to secondarily evaluate other clinical definitions of remission, within an early seropositive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service