Skip to Content
Merck
All Photos(1)

Documents

194654

Sigma-Aldrich

Chrysanthemyl alcohol, mixture of cis and trans

98%

Synonym(s):

2,2-Dimethyl-3-(2-methylpropenyl)cyclopropanemethanol, Chrysanthemol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C3H2[CH=C(CH3)2]CH2OH
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.475 (lit.)

bp

66-69 °C/0.07 mmHg (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\C1C(CO)C1(C)C

InChI

1S/C10H18O/c1-7(2)5-8-9(6-11)10(8,3)4/h5,8-9,11H,6H2,1-4H3

InChI key

HIPIENNKVJCMAP-UHFFFAOYSA-N

General description

Oxidation of chrysanthemyl alcohol in the presence of [RuO4]2- yields chrysantheniic acid.

Application

Chrysanthemyl alcohol was used in the determination of position of double bonds in various terepenes and branched chain compounds.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Attygalle et al.
Analytical chemistry, 65(18), 2528-2533 (1993-09-15)
The electron-impact mass spectra of alpha, beta-bis(methylthio) derivatives of certain terpenes and other compounds with branched alkenyl groups contain diagnostic peaks that can be used for locating the position of the double bond in the parent compound. In contrast to
Oxo complexes of ruthenium (VI) and (VII) as organic oxidants.
Green G, et al.
Journal of the Chemical Society. Perkin Transactions 1, 681-686 (1984)
Gas chromatographic determination of racemic cis- and trans-chrysanthemols and their potential aldehyde and carboxylic acid microbial metabolites.
M Miski et al.
Journal of chromatography, 437(2), 436-441 (1988-03-25)
[Studies on the chemical constituents of Chrysanthemum indicum L].
D Q Yu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 22(11), 837-840 (1987-11-01)
L Y Mou et al.
Journal of Asian natural products research, 3(2), 103-116 (2001-06-16)
Chrysanthemol (1), a trans-eudesmane type sesquiterpene from Chrysanthemum indicum L., possesses certain anti-inflammatory activity. Its total synthesis was approached from two alternative routes and finally accomplished in ten steps from R-(+)-carvone via alpha-eudesmol (10) as the key intermediate. The overall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service