Skip to Content
Merck
All Photos(2)

Key Documents

194115

Sigma-Aldrich

4-Chlorophenyl phenyl sulfone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2C6H5
CAS Number:
Molecular Weight:
252.72
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97%

mp

90-94 °C (lit.)

solubility

acetone: soluble 74.4 g/100ml at 20 °C(lit.)
benzene: soluble 44.4 g /100ml at 20 °C(lit.)
dioxane: soluble 65.6 g/100ml at 20 °C(lit.)
hexane: soluble 0.4 g/100ml at 20 °C(lit.)
isopropanol: soluble 21 g/100ml at 20 °C(lit.)

functional group

chloro
sulfone

SMILES string

Clc1ccc(cc1)S(=O)(=O)c2ccccc2

InChI

1S/C12H9ClO2S/c13-10-6-8-12(9-7-10)16(14,15)11-4-2-1-3-5-11/h1-9H

InChI key

OFCFYWOKHPOXKF-UHFFFAOYSA-N

General description

4-Chlorophenyl phenyl sulfone is also referred to as sulphenone. It participates in palladium-catalyzed amidation reaction in the presence of low CO pressures and an iodide salt[1].

Application

4-Chlorophenyl phenyl sulfone was employed in polyetherification reaction of 4,4′-dihalodiphenyl sulfones[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive dehalogenation versus substitution in the polyetherification of 4, 4'-dihalodiphenyl sulfones with bisphenolates.
Percec V, et al.
Macromolecules, 26(14), 3650-3662 (1993)
Robert J. Perry et al.
The Journal of organic chemistry, 61(21), 7482-7485 (1996-10-18)
The palladium-catalyzed amidation of electron-deficient aryl chlorides proceeds readily in the presence of low CO pressures and a slight excess of an iodide salt. The rates of amidation are accelerated over those without added salt, and iodide is preferred over

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service