Skip to Content
Merck
All Photos(2)

Key Documents

193356

Sigma-Aldrich

3-(Trifluoromethyl)phenylacetic acid

97%

Synonym(s):

(α,α,α-Trifluoro-m-tolyl)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$161.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$161.00

About This Item

Linear Formula:
CF3C6H4CH2CO2H
CAS Number:
Molecular Weight:
204.15
Beilstein:
2213223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

76-79 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)Cc1cccc(c1)C(F)(F)F

InChI

1S/C9H7F3O2/c10-9(11,12)7-3-1-2-6(4-7)5-8(13)14/h1-4H,5H2,(H,13,14)

InChI key

BLXXCCIBGGBDHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trifluoromethyl)phenylacetic acid [(α,α,α-Trifluoro-m-tolyl)acetic acid] was used in the preparation of N,N-diethyl[(α,α,α-triuoro-m-tolyl)]acetamide(DM156)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerome A Klun et al.
Journal of medical entomology, 43(6), 1248-1251 (2006-12-14)
An earlier in vitro screening of N,N-diethyl-3-methylbenzamide (Deet) and N,N-diethylphenylacetamide (DEPA) analogs showed that two DEPA analogs, N,N-diethyl(3-bromophenyl) acetamide and N,N-diethyl[(alpha,alpha,alpha-trifluoro-m-tolyl)]acetamide, and one Deet analog, N,N,diethyl[3-(trifluoromethyl)]benzamide, had biting-deterrent activities that were superior to Deet against Aedes aegypti (L.) and Anopheles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service