Skip to Content
Merck
All Photos(1)

Key Documents

191981

Sigma-Aldrich

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid-co-acrylonitrile)

acrylonitrile ~95 wt. %

Synonym(s):

2-Acrylamido-2-methylpropanesulfonic acid-acrylonitrile copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$333.00

$333.00


Estimated to ship on16 March 2025


Request a Bulk Order

Select a Size

Change View
10 G
$333.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$333.00


Estimated to ship on16 March 2025


Request a Bulk Order

composition

acrylonitrile, ~95 wt. %

Quality Level

SMILES string

C=CC#N.CC(C)(CS(O)(=O)=O)NC(=O)C=C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service