Skip to Content
Merck
All Photos(1)

Key Documents

189170

Sigma-Aldrich

5,6-Dihydro-4-methoxy-2H-pyran

95%

Synonym(s):

4-Methoxy-5,6-dihydropyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
1341205
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

59 °C/20 mmHg (lit.)

density

1.022 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC1=CCOCC1

InChI

1S/C6H10O2/c1-7-6-2-4-8-5-3-6/h2H,3-5H2,1H3

InChI key

FSMHNRHLQAABPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,6-Dihydro-4-methoxy-2H-pyran is suitable reagent for protection of nucleoside hydroxyl functions[1].

Application

5,6-Dihydro-4-methoxy-2H-pyran was used in solid-phase synthesis of series of oligoribonucleotides[2]. It was also used as reagent for the protection of chiral alcohols[3][4].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 1-Diethoxybut-2-ene as a Precursor of (2-Hydroxyethyl)-Substituted Alkoxy Dienes: Convenient Intermediates for a New Synthesis of 2-Substituted and 2, 6-Disubstituted Tetrahydro-4H-pyran-4-ones.
Prandi C and Venturello P.
The Journal of Organic Chemistry, 59(12), 3494-3496 (1994)
C Lehmann et al.
Nucleic acids research, 17(7), 2379-2390 (1989-04-11)
Efficient solid-phase synthesis of a series of oligoribonucleotides of up to 20 residues is described that utilises the 9-fluorenylmethoxycarbonyl group (Fmoc) for 5'-protection and 4-methoxytetrahydropyran-4-yl (Mthp) for 2'-protection of ribonucleotide monomers and a phosphoramidite coupling procedure. The Fmoc group is
A J Leigh et al.
Biochemistry, 31(37), 8978-8983 (1992-09-22)
The substrate stereospecificity of phosphatidylinositol-specific phospholipase C from Bacillus cereus is examined using the resolved optical isomers of synthetic myo-inositol 1-(4-nitrophenyl phosphate), a chromogenic substrate for the phospholipase. The synthetic route employs mild acid-labile protecting groups and separation of the
Tetrahedron Letters, 33, 2371-2371 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service