Skip to Content
Merck
All Photos(1)

Key Documents

188190

Sigma-Aldrich

o-Cresolphthalein Complexone

Synonym(s):

o-Cresolphthalein-3′,3′′-bis-methyleneiminodiacetic acid, o-Cresolphthalexon, Metal phthalein, Phthalein purple, Xylenylphthalein-bisiminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32N2O12
CAS Number:
Molecular Weight:
636.60
Beilstein:
381994
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

technique(s)

titration: suitable

mp

181-185 °C (dec.) (lit.)

λmax

377 nm (2nd)
575 nm

SMILES string

Cc1cc(cc(CN(CC(O)=O)CC(O)=O)c1O)C2(OC(=O)c3ccccc23)c4cc(C)c(O)c(CN(CC(O)=O)CC(O)=O)c4

InChI

1S/C32H32N2O12/c1-17-7-21(9-19(29(17)43)11-33(13-25(35)36)14-26(37)38)32(24-6-4-3-5-23(24)31(45)46-32)22-8-18(2)30(44)20(10-22)12-34(15-27(39)40)16-28(41)42/h3-10,43-44H,11-16H2,1-2H3,(H,35,36)(H,37,38)(H,39,40)(H,41,42)

InChI key

IYZPEGVSBUNMBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Corns
Annals of clinical biochemistry, 27 ( Pt 2), 152-155 (1990-03-01)
The spectral interference caused by haemoglobin in the calcium-cresolphthalein complexone reaction was investigated. The results show that the nature of the haemoglobin derivative in the reaction mixture influences the degree of interference; in turn, the derivative formed depends on the
B Aghabeigi et al.
International journal of oral and maxillofacial surgery, 31(6), 646-649 (2003-01-11)
Saline aspirates have been commonly used in the biochemical investigations of temporomandibular joint (TMJ) pathology. However, due to presence of adhesions in the diseased temporomandibular joint, full equilibration between the injected saline and the synovial fluid may not be achieved
D H Patterson et al.
Journal of chromatography. A, 662(2), 389-395 (1994-02-25)
Electrophoretically mediated microanalysis (EMMA) was used for the determination of calcium. The faster analyte zone containing the calcium was injected spatially behind a slower zone of o-cresolphthalein complexone in a capillary electrophoresis based system. Upon application of the electric field
James H Wible et al.
Radiology, 233(1), 158-164 (2004-09-30)
To measure serum calcium concentration with three different analytic methods after administration of gadoversetamide and three other gadolinium chelates in dogs. Six dogs were injected with 0.1-, 0.3-, 1.0-, and 3.0-mmol/kg doses of gadoversetamide; 1.0-mmol/kg doses of gadodiamide, gadopentetate, and
J Toffaletti et al.
Clinical chemistry, 26(11), 1562-1565 (1980-10-01)
We adapted a method for dialyzable calcium from fluorometric detection by use of calcein to a more specific spectrophotometric determination with cresolphthalein complexone. The reagents are available commercially and perform satisfactorily with respect to noise level, drift, stability, and sensitivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service