Skip to Content
Merck
All Photos(1)

Key Documents

188107

Sigma-Aldrich

2-Naphthaleneethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CH2OH
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

180-184 °C/15 mmHg (lit.)

functional group

hydroxyl

SMILES string

OCCc1ccc2ccccc2c1

InChI

1S/C12H12O/c13-8-7-10-5-6-11-3-1-2-4-12(11)9-10/h1-6,9,13H,7-8H2

InChI key

VCZANYLMPFRUHG-UHFFFAOYSA-N

Gene Information

human ... BAD(572)

General description

2-Naphthaleneethanol undergoes esterification reaction with poly(ethylene glycol) monomethyl ether carboxylic acid[1]. Emission spectra of 2-naphthaleneethanol reacted with UV-irradiated octadecylsiloxane self-assembled monolayers has been investigated[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Nagaoka et al.
Bioscience, biotechnology, and biochemistry, 64(4), 781-784 (2000-06-01)
Kinetic resolution of racemic alcohols, (+/-)-1-(4-substituted phenyl)ethanol and (+/-)-1-(2-naphthyl)ethanol, was done with immobilized green pea, soybean, or buckwheat proteins. The resolution was done stereoselectively by oxidizing only one enantiomer of a racemic alcohol to leave an optically active alcohol with
Eric A McArthur et al.
Journal of the American Chemical Society, 126(8), 2260-2261 (2004-02-26)
Detection and quantification of submonolayer coverage surface species is not trivial. We have developed a novel method sensitive to surface-bound chemical functional groups as low as 10(11) molecules/cm(2) by specific covalent attachment of fluorescent chromophores. This enables the intermediates of
V Schurig et al.
Journal of biochemical and biophysical methods, 43(1-3), 223-240 (2000-06-28)
The use of complexation SFC for enantiomer separation of Lewis base selectands on chiral nickel(II)- and zinc(II)-bis[(3-heptafluorobutanoyl)-10-methylene-(1R)-camphora te] chemically bonded to poly(dimethylsiloxane) (Chirasil-nickel and Chirasil-zinc) and employed as Lewis acid selectors is described. The method is especially suited for less
N Seurre et al.
Physical chemistry chemical physics : PCCP, 8(8), 1007-1016 (2006-02-17)
Jet-cooled diastereoisomeric complexes formed between a chiral probe, (+/-)-2-naphthyl-1-ethanol, and chiral lactic acid derivatives have been characterised by laser-induced fluorescence and IR fluorescence-dip spectroscopy. Complexes with non chiral alpha-hydroxyesters and chiral beta-hydroxyesters have also been studied for the sake of
H Nagaoka et al.
Bioscience, biotechnology, and biochemistry, 63(11), 1991-1992 (2000-01-15)
(S)-1-(2-Naphthyl)ethanol was yielded by immobilized pea (Pisum sativum L.) protein (IPP) from (R, S) 2-naphthyl ethanol (> 99% ee, yield; about 50%), in which the (R)-enantiomer was selectively oxidized to 2-acetonaphthone. IPP could be reused consecutively at least three times

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service