Skip to Content
Merck
All Photos(1)

Key Documents

187720

Sigma-Aldrich

2-Chloroanthracene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl
CAS Number:
Molecular Weight:
212.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

221-223 °C (lit.)

SMILES string

Clc1ccc2cc3ccccc3cc2c1

InChI

1S/C14H9Cl/c15-14-6-5-12-7-10-3-1-2-4-11(10)8-13(12)9-14/h1-9H

InChI key

OWFINXQLBMJDJQ-UHFFFAOYSA-N

Application

2-Chloroanthracene was employed as model compound to investigate the formation of polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans in fly ash[1]. It was also used in the synthesis of polyfluorenes having anthraquinone or naphtylimide end caps[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of PCDDs and PCDFs from anthracene and chloroanthracene in a model fly ash system.
M H Schoonenboom et al.
Environmental science & technology, 29(8), 2005-2009 (1995-08-01)
Sadayuki Asaoka et al.
Journal of the American Chemical Society, 130(36), 11912-11920 (2008-08-14)
Polyfluorenes (pF) were synthesized having anthraquinone (AQ) or naphtylimide (NI) end caps that trap electrons or di- p-tolylaminophenyl (APT2) caps that trap holes. The average lengths of the pF chains in these molecules varied from 7 to 30 nm. End
Yao Zhou et al.
Toxicology in vitro : an international journal published in association with BIBRA, 72, 105098-105098 (2021-01-22)
Chlorinated paraffins (CPs) are produced at more than one million tons per year. Technical CPs mixtures may contain impurities, which end up in consumer products. In the present study, 17 technical CPs mixtures were investigated for the potential occurrence of
Rong Jin et al.
Journal of chromatography. A, 1509, 114-122 (2017-06-18)
Isotopic dilution gas chromatography combined with high resolution mass spectrometry (GC/HRMS) has overwhelming advantages with respect to the accuracy of congener-specific ultratrace analysis of complex persistent organic pollutants (POPs) in environmental matrices. However, an isotopic dilution GC/HRMS method for analysis
Jian-Lin Sun et al.
Ecotoxicology and environmental safety, 113, 31-37 (2014-12-09)
Halogenated polycyclic aromatic hydrocarbons (HPAHs) have been reported to occur widely in urban air. Nevertheless, knowledge about the human health risk associated with inhalation exposure to HPAHs is scarce so far. In the present study, nine HPAHs and 16 PAHs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service