Skip to Content
Merck
All Photos(1)

Key Documents

186996

Sigma-Aldrich

5-Ethyl-2-furaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$72.90

$72.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$72.90

About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$72.90


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.522 (lit.)

bp

79-81 °C/12 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(CC)o1

InChI

1S/C7H8O2/c1-2-6-3-4-7(5-8)9-6/h3-5H,2H2,1H3

InChI key

XADGZBXFWQHBDB-UHFFFAOYSA-N

Application

5-Ethyl-2-furaldehyde was used in the synthesis of N-(tert-butoxycarbonyl)-3-(benzoxazol-5-yl)alanine methyl ester derivatives[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New fluorescent probes based on 3-(2-benzoxazol-5-yl) alanine skeleton-Synthesis and photophysical properties.
Guzow K, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 187(1), 87-96 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service