Skip to Content
Merck
All Photos(1)

Key Documents

186678

Sigma-Aldrich

2,4,4-Trimethyl-1-pentanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

168-169 °C/740 mmHg (lit.)

density

0.818 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(CO)CC(C)(C)C

InChI

1S/C8H18O/c1-7(6-9)5-8(2,3)4/h7,9H,5-6H2,1-4H3

InChI key

ZNRVRWHPZZOTIE-UHFFFAOYSA-N

General description

2,4,4-Trimethyl-1-pentanol is the principal metabolite of 2,2,4-trimethylpentane[1]. Phase equilibria of 2,4,4-trimethyl-1-pentanol in supercritical carbon dioxide has been investigated[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase equilibria of alcohols in supercritical fluids Part II. The effect of side branching on C8 alcohols in supercritical carbon dioxide.
Schwarz CE, et al.
Journal of Supercritical Fluids, 51(2), 128-135 (2009)
C T Olson et al.
Biochemical and biophysical research communications, 130(1), 313-316 (1985-07-16)
The compound 2,2,4-trimethylpentane (2,2,4 TMP) is reported to be especially potent in inducing kidney lesions in male rats (1,2). Although the pathology produced by 2,2,4 TMP has been examined (1), there are no reports concerning the metabolism of 2,2,4 TMP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service