Skip to Content
Merck
All Photos(1)

Key Documents

185663

Sigma-Aldrich

5-Fluoro-1-indanone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$191.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$191.00

About This Item

Empirical Formula (Hill Notation):
C9H7FO
CAS Number:
Molecular Weight:
150.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$191.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

113-114 °C (lit.)

mp

38-40 °C (lit.)

density

1.216 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

Fc1ccc2C(=O)CCc2c1

InChI

1S/C9H7FO/c10-7-2-3-8-6(5-7)1-4-9(8)11/h2-3,5H,1,4H2

InChI key

WVPPBVAMKNQXJA-UHFFFAOYSA-N

Application

5-Fluoro-1-indanone was used to prepare precursor for thiosemicarbazones derived from 1-indanones[1]. It was used in the synthesis of 6-fluoro-1,2,3,4-tetrahydroquinoline hydrochloride[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María E Caputto et al.
Bioorganic & medicinal chemistry, 19(22), 6818-6826 (2011-10-18)
In the present work, we synthesized a series of thiosemicarbazones derived from 1-indanones with good anti-Trypanosoma cruzi activity. Most of them displayed remarkable trypanosomicidal activity. All the compounds showed nonspecific cytotoxicity on human erythrocytes. The ability of the new compounds
Satoshi Sakami et al.
Bioorganic & medicinal chemistry, 16(17), 7956-7967 (2008-08-15)
We have previously reported on antitussive effect of (5R,9R,13S,14S)-17-cyclopropylmethyl-6,7-didehydro-4,5-epoxy-5',6'-dihydro-3-methoxy-4'H-pyrrolo[3,2,1-ij]quinolino[2',1':6,7]morphinan-14-ol(1b) methanesulfonate (TRK-850), a selective delta opioid receptor antagonist which markedly reduced the number of coughs in a rat cough model. We designed TRK-850 based on naltrindole (NTI), a typical delta opioid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service