Skip to Content
Merck
All Photos(1)

Key Documents

18486

Sigma-Aldrich

3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl isocyanate

≥97.0%

Synonym(s):

1H,1H,2H,2H-Perfluorodecyl isocyanate, 2-(Perfluorooctyl)ethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H4F17NO
CAS Number:
Molecular Weight:
489.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥97.0%

form

solid

bp

65 °C/0.5 mmHg

mp

26-31 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCN=C=O

InChI

1S/C11H4F17NO/c12-4(13,1-2-29-3-30)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)10(24,25)11(26,27)28/h1-2H2

InChI key

XBIIIDIAZYSZBI-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Liver, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service