Skip to Content
Merck
All Photos(1)

Key Documents

184330

Sigma-Aldrich

4-Fluorophenyl methyl sulfone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2CH3
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

78-81 °C (lit.)

functional group

fluoro
sulfone

SMILES string

CS(=O)(=O)c1ccc(F)cc1

InChI

1S/C7H7FO2S/c1-11(9,10)7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

DPJHZJGAGIWXTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluorophenyl methyl sulfone was used in the preparation of 4-(4′-trifluoromethoxyphenoxy)phenyl methyl sulfone[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol K Wada et al.
Journal of medicinal chemistry, 45(1), 219-232 (2002-01-05)
A novel series of sulfone N-formylhydroxylamines (retrohydroxamates) have been investigated as matrix metalloproteinases (MMP) inhibitors. The substitution of the ether linkage of ABT-770 (5) with a sulfone group 13a led to a substantial increase in activity against MMP-9 but was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service