Skip to Content
Merck
All Photos(1)

Documents

184209

Sigma-Aldrich

1-Adamantanemethanol

99%

Synonym(s):

1-(Hydroxymethyl)adamantane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
Beilstein:
1853339
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

114-117 °C (lit.)

SMILES string

OCC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C11H18O/c12-7-11-4-8-1-9(5-11)3-10(2-8)6-11/h8-10,12H,1-7H2/t8-,9+,10-,11-

InChI key

MDVGOOIANLZFCP-BIBSGERRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Adamantanemethanol acts as guest molecule and forms β-cyclodextrin complexes. It reacts with 3,4,5,6-tetrafluorophthalonitrile to yield 3,4,5,6-tetra-(1-adamantylmethoxy)phthalonitrile.

Application

1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Chyba et al.
Inorganic chemistry, 57(15), 8735-8747 (2018-04-06)
The potential of paramagnetic ruthenium(III) compounds for use as anticancer metallodrugs has been investigated extensively during the past several decades. However, the means by which these ruthenium compounds are transported and distributed in living bodies remain relatively unexplored. In this
A S Kostense et al.
Journal of computer-aided molecular design, 5(6), 525-543 (1991-12-01)
A series of beta-cyclodextrin complexes containing various guest molecules was studied using computer-aided molecular modeling and conformation analysis techniques. The geometry of each complex was studied using crystallographic data. The positions of the glycosidic O4 atoms indicate that the beta-cyclodextrin
Syntheses and characterization of phthalonitriles and phthalocyanines substituted with adamantane moieties.
Causey PW, et al.
Canadian Journal of Chemistry, 84(10), 1380-1387 (2006)
Sanlei Xie et al.
Analytical and bioanalytical chemistry (2018-07-15)
A sensitive competitive immunoassay with simple operation was developed for the detection of the anti-virus drug amantadine (AMD). The single-chain variable fragment (scFv) antibody against AMD was site-specific biotinylated and overexpressed as a secreted body in Escherichia coli AVB101. Horseradish
Highly photostable silicon (IV) phthalocyanines containing adamantane moieties: synthesis, structure, and properties.
Shen X-M, et al.
Tetrahedron, 66(46), 9041-9048 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service