Skip to Content
Merck
All Photos(1)

Documents

183539

Sigma-Aldrich

5-Methoxy-1-indanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
1282137
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

107-109 °C (lit.)

SMILES string

COc1ccc2C(=O)CCc2c1

InChI

1S/C10H10O2/c1-12-8-3-4-9-7(6-8)2-5-10(9)11/h3-4,6H,2,5H2,1H3

InChI key

QOPRWBRNMPANKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The cytotoxic potential of 5-methoxy-1-indanone was assessed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiffany J Somers-Edgar et al.
Anti-cancer drugs, 20(1), 33-40 (2008-10-03)
We postulated that methoxy-substituted cyclic compounds could inhibit estrogen receptor (ER) negative breast cancer growth in vitro. Therefore, this study assessed the cytotoxic potential of various methoxy-substituted cyclic compounds [7,8-dimethoxyflavone, 4-methoxyphenylacetic acid, 2-methoxyphenylacetic acid, 4-methoxybenzophenone, 5-methoxy-1-indanone, and coenzyme Q0 (CoQ0)]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service